siRNA Details
| Gene Symbol: Srp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009273.4 | NP_033299.1 | 5 | 446 | ||
| GenBank mRNA | AK019310.1 | - | 5 | 429 | ||
| AK156845.1 | - | 386 | ||||
| AK167821.1 | - | 5 | 395 | |||
| AK168144.1 | - | 5 | 414 | |||
| BC021537.1 | - | 5 | 422 | 5353575 | ||
| M29264.1 | - | 5 | 371 | |||

Srp14
Genomic Map