siRNA Details
| Gene Symbol: Srprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009275.4 | NP_033301.1 | 7 | 1076 | ||
| GenBank mRNA | AK004854.1 | - | 7 | 1109 | ||
| AK008383.1 | - | 7 | 1119 | |||
| AK015469.1 | - | 7 | 1141 | |||
| AK030437.1 | - | 7 | 1077 | |||
| AK075583.1 | - | 7 | 1079 | |||
| AK160600.1 | - | 6 | 2310 | |||
| AK167039.1 | - | 7 | 1082 | |||
| AK182239.1 | - | 1 | 66 | |||
| AK208005.1 | - | 1 | 36 | |||
| BC003798.1 | - | 7 | 1095 | 3490766 | ||

Srprb
Genomic Map