siRNA Details
| Gene Symbol: Trim21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082552.2 | NP_001076021.1 | 6 | 900 | ||
| NM_009277.4 | NP_033303.3 | 7 | 969 | |||
| GenBank mRNA | AK141294.1 | - | 7 | 949 | ||
| AK150484.1 | - | 6 | 864 | |||
| AK152619.1 | - | 6 | 864 | |||
| AK171489.1 | - | 7 | 934 | |||
| BC010580.1 | - | 7 | 909 | 3584654 | ||
| CT010336.1 | - | 5 | 794 | |||
| L27990.1 | - | 7 | 934 | |||

Trim21
Genomic Map