siRNA Details
Gene Symbol: Zfp143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009281.3 | NP_033307.2 | 12 | 1332 | ||
GenBank mRNA | AF011758.2 | - | 1281 | |||
AK028056.1 | - | 12 | 1325 | |||
AK029078.1 | - | 12 | 1324 | |||
AK029926.1 | - | 12 | 1597 | |||
BC037658.1 | - | 11 | 1242 | 5356405 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
