siRNA Details
| Gene Symbol: Stim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009287.4 | NP_033313.2 | 3 | 919 | ||
| GenBank mRNA | AK041944.1 | - | 3 | 938 | ||
| AK086594.1 | - | 3 | 926 | |||
| AK201690.1 | - | 3 | 220 | |||
| BC021644.1 | - | 3 | 921 | 4191156 | ||
| U47323.1 | - | 3 | 521 | |||

Stim1
Genomic Map