siRNA Details
| Gene Symbol: Slk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164639.1 | NP_001158111.1 | 15 | 3387 | ||
| NM_009289.3 | NP_033315.2 | 16 | 3480 | |||
| GenBank mRNA | AF039574.1 | - | 16 | 3485 | ||
| AF112855.1 | - | 15 | 3276 | |||
| AK037798.1 | - | 7 | 1361 | |||
| AK122218.1 | - | 15 | 3728 | |||
| BC131674.1 | - | 15 | 3288 | 40110915 | ||
| BC131675.1 | - | 16 | 3381 | 40110921 | ||
| CO431234.1 | - | 2 | 268 | |||

Slk
Genomic Map