siRNA Details
| Gene Symbol: Stra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009292.1 | NP_033318.1 | 5, 6 | 749 | ||
| GenBank mRNA | AK006887.1 | - | 6, 7 | 1090 | ||
| BC100746.1 | - | 4, 5 | 497 | |||
| BC103590.1 | - | 5, 6 | 695 | 40045824 | ||
| BC103591.1 | - | 5, 6 | 695 | 40045823 | ||
| BC103603.1 | - | 5, 6 | 692 | |||
| Z75287.1 | - | 5, 6 | 749 | |||

Stra8
Genomic Map