siRNA Details
| Gene Symbol: Supt4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009296.1 | NP_033322.1 | 5 | 519 | ||
| GenBank mRNA | AK002990.1 | - | 5 | 621 | ||
| AK146235.1 | - | 6 | 676 | |||
| AK167941.1 | - | 5 | 533 | |||
| AK168229.1 | - | 6 | 676 | |||
| BC024391.1 | - | 5 | 514 | 5011064 | ||
| BC061174.1 | - | 5 | 520 | 6592996 | ||
| BC087923.1 | - | 5 | 495 | 30308013 | ||
| U43154.1 | - | 5 | 485 | |||

Supt4a
Genomic Map