siRNA Details
| Gene Symbol: Sva | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009299.2 | NP_033325.2 | 4 | 448 | ||
| GenBank mRNA | BC104304.1 | - | 4 | 446 | 40043712 | |
| BC104305.1 | - | 4 | 448 | 40043713 | ||
| CJ055470.1 | - | 4 | 448 | |||
| M94179.1 | - | 4 | 434 | |||

Sva
Genomic Map