siRNA Details
| Gene Symbol: Swap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009302.3 | NP_033328.3 | 2 | 248 | ||
| GenBank mRNA | AF053974.1 | - | 2 | 140 | ||
| AK019882.1 | - | 2 | 191 | |||
| AK041505.1 | - | 2 | 196 | |||
| AK045295.1 | - | 2 | 220 | |||
| AK146773.1 | - | 2 | 240 | |||
| AK172990.1 | - | 2 | 185 | |||
| BC065136.1 | - | 2 | 190 | 30546814 | ||
| BY118458.1 | - | 2 | 249 | |||

Swap70
Genomic Map