siRNA Details
| Gene Symbol: Swap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009302.3 | NP_033328.3 | 11, 12 | 1767 | ||
| GenBank mRNA | AF053974.1 | - | 11, 12 | 1659 | ||
| AK019882.1 | - | 11, 12 | 1711 | |||
| AK146773.1 | - | 11, 12 | 1759 | |||
| AK150621.1 | - | 5, 6 | 739 | |||
| AK163357.1 | - | 4, 5 | 577 | |||
| AK172990.1 | - | 11, 12 | 1704 | |||
| BC065136.1 | - | 11, 12 | 1709 | 30546814 | ||

Swap70
Genomic Map