siRNA Details
| Gene Symbol: Pou6f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010127.3 | NP_034257.1 | 6 | 3696 | ||
| GenBank mRNA | AK009674.1 | - | 1 | 1121 | ||
| BC053046.1 | - | 8 | 3526 | |||
| BC059830.1 | - | 6 | 3696 | 6410550 | ||
| BC085139.1 | - | 11 | 4074 | 6406821 | ||

Pou6f1
Genomic Map