siRNA Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011101.3 | NP_035231.2 | 5, 6 | 756 | ||
| GenBank mRNA | AK147084.1 | - | 5, 6 | 757 | ||
| BC096493.1 | - | 5, 6 | 550 | 30742571 | ||
| M25811.1 | - | 5, 6 | 790 | |||
| X52684.1 | - | 6, 7 | 697 | |||
| X52685.1 | - | 6, 7 | 697 | |||

Prkca
Genomic Map