siRNA Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011101.3 | NP_035231.2 | 9 | 1204 | ||
| GenBank mRNA | AK147084.1 | - | 9 | 1205 | ||
| BC096493.1 | - | 9 | 998 | 30742571 | ||
| M25811.1 | - | 9 | 1238 | |||
| X52684.1 | - | 10 | 1145 | |||
| X52685.1 | - | 10 | 1145 | |||

Prkca
Genomic Map