siRNA Details
| Gene Symbol: Pld3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011116.1 | NP_035246.1 | 10 | 1231 | ||
| GenBank mRNA | AF026124.1 | - | 10 | 1231 | ||
| AK153508.1 | - | 8 | 920 | |||
| BC068170.1 | - | 10 | 1227 | |||
| BC076586.1 | - | 10 | 1295 | 30604186 | ||

Pld3
Genomic Map