siRNA Details
Gene Symbol: Pole2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011133.2 | NP_035263.1 | 9, 10 | 716 | ||
GenBank mRNA | AF036898.1 | - | 9, 10 | 689 | ||
AK077592.1 | - | 9, 10 | 717 | |||
AK172024.1 | - | 9, 10 | 711 | |||
BC052170.1 | - | 9, 10 | 705 | |||
BC063772.1 | - | 9, 10 | 705 | 30147626 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
