siRNA Details
| Gene Symbol: Ppef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011148.3 | NP_035278.1 | 3, 4 | 238 | ||
| GenBank mRNA | AF023458.1 | - | 3, 4 | 238 | ||
| BC027049.1 | - | 4, 5 | 545 | 5356685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppef2| Gene Symbol: Ppef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011148.3 | NP_035278.1 | 3, 4 | 238 | ||
| GenBank mRNA | AF023458.1 | - | 3, 4 | 238 | ||
| BC027049.1 | - | 4, 5 | 545 | 5356685 | ||