siRNA Details
| Gene Symbol: Prkg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013833.3 | NP_001013855.1 | 3 | 723 | ||
| NM_011160.3 | NP_035290.1 | 3 | 646 | |||
| GenBank mRNA | AF084547.1 | - | 3 | 646 | ||
| AK083991.1 | - | 3 | 723 | |||
| BC113162.1 | - | 3 | 628 | 40060049 | ||

Prkg1
Genomic Map