siRNA Details
| Gene Symbol: Mapk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011161.5 | NP_035291.4 | 12 | 1528 | ||
| GenBank mRNA | AK144846.1 | - | 12 | 1501 | ||
| AK161190.1 | - | 10 | 1703 | |||
| AK161562.1 | - | 12 | 1480 | |||
| BC064737.1 | - | 12 | 1477 | 30112690 | ||
| BC092526.1 | - | 12 | 1528 | 6593321 | ||

Mapk11
Genomic Map