siRNA Details
| Gene Symbol: Psmb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011185.3 | NP_035315.1 | 4 | 331 | ||
| GenBank mRNA | AK028265.1 | - | 4 | 352 | ||
| AK077520.1 | - | 4 | 332 | |||
| AK132161.1 | - | 4 | 332 | |||
| AK167647.1 | - | 4 | 353 | |||
| BC018351.1 | - | 4 | 332 | 3584538 | ||
| U60824.1 | - | 4 | 346 | |||
| X80686.1 | - | 4 | 335 | |||

Psmb1
Genomic Map