siRNA Details
| Gene Symbol: Ptpn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011206.2 | NP_035336.2 | 10, 11 | 923 | ||
| GenBank mRNA | AK144353.1 | - | 10, 11 | 905 | ||
| BC008512.1 | - | 10, 11 | 893 | 3591418 | ||
| U35124.1 | - | 10, 11 | 897 | |||
| U49853.1 | - | 10, 11 | 863 | |||
| U52523.1 | - | 10, 11 | 911 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001013111, BC093398, BC098788.

Ptpn18
Genomic Map