siRNA Details
| Gene Symbol: Rad17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044371.2 | NP_001037836.1 | 4, 5 | 554 | ||
| NM_011233.3 | NP_035363.2 | 4, 5 | 682 | |||
| GenBank mRNA | AF085737.1 | - | 4, 5 | 605 | ||
| AJ011923.1 | - | 3, 4 | 455 | |||
| AK134277.1 | - | 4, 5 | 509 | |||
| AK162016.1 | - | 4, 5 | 541 | |||
| BC023939.1 | - | 4, 5 | 559 | |||
| BC050267.1 | - | 1, 2 | 194 | |||
| BC066855.1 | - | 4, 5 | 672 | 30473984 | ||

Rad17
Genomic Map