siRNA Details
| Gene Symbol: Rad17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044371.2 | NP_001037836.1 | 6 | 821 | ||
| NM_001283011.1 | NP_001269940.1 | 7 | 1026 | |||
| NM_011233.3 | NP_035363.2 | 6 | 949 | |||
| GenBank mRNA | AF085737.1 | - | 6 | 872 | ||
| AJ011923.1 | - | 5 | 722 | |||
| AK134277.1 | - | 6 | 776 | |||
| AK162016.1 | - | 6 | 808 | |||
| AK167358.1 | - | 7 | 969 | |||
| BC023939.1 | - | 6 | 826 | |||

Rad17
Genomic Map