siRNA Details
| Gene Symbol: SLC16A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174166.1 | NP_001167637.1 | 7 | 2342 | ||
| NM_004694.4 | NP_004685.2 | 6 | 2215 | |||
| XM_005257789.3 | XP_005257846.2 | 6 | 2380 | |||
| XM_011525461.2 | XP_011523763.1 | 6 | 2059 | |||
| XM_017025291.1 | XP_016880780.1 | 7 | 2179 | |||
| XM_017025292.1 | XP_016880781.1 | 6 | 2242 | |||
| GenBank mRNA | AX750581.1 | - | 7 | 2194 | ||
| BC064832.1 | - | 6 | 2217 | 6498761 | ||
| U79745.1 | - | 7 | 2194 | |||

SLC16A6
Genomic Map