siRNA Details
| Gene Symbol: SLC16A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271765.1 | NP_001258694.1 | 7 | 1979 | ||
| NM_004695.3 | NP_004686.1 | 7 | 1913 | |||
| XM_005257790.1 | XP_005257847.1 | 6 | 1855 | |||
| XM_011525462.1 | XP_011523764.1 | 6 | 1803 | |||
| XM_017025293.1 | XP_016880782.1 | 6 | 3640 | |||
| GenBank mRNA | AK000416.1 | - | 7 | 1979 | ||
| AK092512.1 | - | 6 | 2022 | |||
| BC009684.1 | - | 7 | 1894 | 3895881 | ||
| BC033611.1 | - | 6 | 1803 | 5218712 | ||
| U59299.1 | - | 5 | 1667 | |||

SLC16A5
Genomic Map