siRNA Details
Gene Symbol: Ros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011282.2 | NP_035412.2 | 21 | 3311 | ||
GenBank mRNA | U15443.1 | - | 21 | 3310 | ||
X81650.1 | - | 21 | 3134 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011282.2 | NP_035412.2 | 21 | 3311 | ||
GenBank mRNA | U15443.1 | - | 21 | 3310 | ||
X81650.1 | - | 21 | 3134 |