siRNA Details
| Gene Symbol: Rp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195662.1 | NP_001182591.1 | 2 | 198 | ||
| NM_011283.2 | NP_035413.1 | 2 | 241 | |||
| GenBank mRNA | AF155141.2 | - | 2 | 232 | ||
| AK044413.1 | - | 2 | 234 | |||
| AK141793.1 | - | 2 | 199 | |||
| BC038794.1 | - | 2 | 181 | |||
| BC096607.1 | - | 2 | 241 | |||
| BC120927.1 | - | 2 | 195 | 40099705 | ||
| BC120928.1 | - | 2 | 195 | 40099706 | ||

Rp1
Genomic Map