siRNA Details
| Gene Symbol: Scn8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077499.2 | NP_001070967.1 | 5 | 702 | ||
| NM_011323.3 | NP_035453.2 | 4 | 603 | |||
| GenBank mRNA | AF049617.1 | - | 4 | 549 | ||
| AK158609.1 | - | 5 | 664 | |||
| U26707.1 | - | 4 | 561 | |||

Scn8a
Genomic Map