siRNA Details
| Gene Symbol: SCAF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004719.2 | NP_004710.2 | 11 | 1343 | ||
| XM_005269230.2 | XP_005269287.2 | 11 | 1233 | |||
| XM_011538984.2 | XP_011537286.1 | 12 | 2862 | |||
| XM_011538985.1 | XP_011537287.1 | 11 | 1177 | |||
| XM_017020217.1 | XP_016875706.1 | 12 | 1221 | |||
| XM_017020218.1 | XP_016875707.1 | 12 | 1221 | |||
| XM_017020219.1 | XP_016875708.1 | 12 | 1420 | |||
| XM_017020220.1 | XP_016875709.1 | 12 | 1221 | |||
| XM_017020221.1 | XP_016875710.1 | 10 | 1550 | |||
| XM_017020222.1 | XP_016875711.1 | 9 | 1520 | |||
| XM_017020223.1 | XP_016875712.1 | 12 | 1443 | |||
| GenBank mRNA | AF030234.1 | - | 11 | 1291 | ||
| AK026473.1 | - | 12 | 1343 | |||
| AK302999.1 | - | 11 | 1268 | |||
| AK314093.1 | - | 5 | 606 | |||
| BC017465.2 | - | 8 | 792 | |||
| BC033589.1 | - | 7 | 748 | |||
| BC040951.2 | - | 3 | 424 | |||
| Y11251.1 | - | 1 | 1321 | |||

SCAF11
Genomic Map