siRNA Details
| Gene Symbol: Sema3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011348.2 | NP_035478.2 | 9, 10 | 1640 | ||
| GenBank mRNA | AF034744.1 | - | 9, 10 | 1375 | ||
| AK129116.1 | - | 4, 5 | 405 | |||
| AK141295.1 | - | 8, 9 | 2362 | |||
| AK142340.1 | - | 9, 10 | 1467 | |||
| AK159214.1 | - | 9, 10 | 1431 | |||
| BC057956.1 | - | 9, 10 | 1538 | 5357516 | ||
| Z80941.1 | - | 9, 10 | 1006 | |||
| Z93947.1 | - | 9, 10 | 1615 | |||
| Z93948.1 | - | 9, 10 | 1615 | |||

Sema3e
Genomic Map