siRNA Details
| Gene Symbol: Shbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011367.2 | NP_035497.1 | 2 | 216 | ||
| GenBank mRNA | BC120785.1 | - | 2 | 201 | 40129708 | |
| BC120787.1 | - | 2 | 201 | 40129710 | ||
| U85644.1 | - | 2 | 179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shbg| Gene Symbol: Shbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011367.2 | NP_035497.1 | 2 | 216 | ||
| GenBank mRNA | BC120785.1 | - | 2 | 201 | 40129708 | |
| BC120787.1 | - | 2 | 201 | 40129710 | ||
| U85644.1 | - | 2 | 179 | |||