siRNA Details
| Gene Symbol: Shcbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011369.2 | NP_035499.1 | 3 | 310 | ||
| GenBank mRNA | AF017152.1 | - | 3 | 310 | ||
| AK052944.1 | - | 2 | 459 | |||
| AK137496.1 | - | 3 | 326 | |||
| AK144966.1 | - | 3 | 333 | |||
| AK149580.1 | - | 3 | 327 | |||
| AK162943.1 | - | 3 | 324 | |||
| BC070455.1 | - | 3 | 330 | 30606785 | ||

Shcbp1
Genomic Map