siRNA Details
| Gene Symbol: Shcbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011369.2 | NP_035499.1 | 2 | 224 | ||
| GenBank mRNA | AF017152.1 | - | 2 | 224 | ||
| AK051205.1 | - | 2 | 258 | |||
| AK052944.1 | - | 2 | 250 | |||
| AK137496.1 | - | 2 | 240 | |||
| AK144966.1 | - | 2 | 247 | |||
| AK149580.1 | - | 2 | 241 | |||
| AK162943.1 | - | 2 | 238 | |||
| BC070455.1 | - | 2 | 244 | 30606785 | ||

Shcbp1
Genomic Map