siRNA Details
| Gene Symbol: St6galnac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011372.2 | NP_035502.1 | 5 | 1269 | ||
| GenBank mRNA | AK034804.1 | - | 5 | 1271 | ||
| BC058387.1 | - | 5 | 1254 | 6817021 | ||
| Y11342.1 | - | 5 | 1268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St6galnac3| Gene Symbol: St6galnac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011372.2 | NP_035502.1 | 5 | 1269 | ||
| GenBank mRNA | AK034804.1 | - | 5 | 1271 | ||
| BC058387.1 | - | 5 | 1254 | 6817021 | ||
| Y11342.1 | - | 5 | 1268 | |||