siRNA Details
| Gene Symbol: Slc8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112798.2 | NP_001106269.1 | 2 | 1138 | ||
| NM_001286684.1 | NP_001273613.1 | 2 | 1138 | |||
| NM_011406.3 | NP_035536.2 | 2 | 1138 | |||
| GenBank mRNA | AB080744.1 | - | 1 | 207 | ||
| AF004666.1 | - | 2 | 1133 | |||
| AK048160.1 | - | 705 | ||||
| BC079673.1 | - | 1 | 1029 | 30532156 | ||
| DQ388998.1 | - | 1 | 1028 | |||
| U70033.1 | - | 2 | 1128 | |||

Slc8a1
Genomic Map