siRNA Details
| Gene Symbol: Sp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018042.3 | NP_001018052.1 | 4 | 487 | ||
| NM_001098425.1 | NP_001091895.1 | 2 | 396 | |||
| GenBank mRNA | AF062567.1 | - | 2 | 123 | ||
| AK004607.1 | - | 1 | 137 | |||
| AK088238.1 | - | 2 | 371 | |||
| AK132702.1 | - | 2 | 142 | |||
| BC079874.1 | - | 4 | 487 | 6833706 | ||
| CA328978.1 | - | 2 | 400 | |||
| CJ167744.1 | - | 2 | 397 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008761975, AA875516.

Sp3
Genomic Map