siRNA Details
| Gene Symbol: Serpinb9c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164524.1 | NP_001157996.1 | 7 | 857 | ||
| NM_011453.6 | NP_035583.1 | 7 | 828 | |||
| GenBank mRNA | AK014448.1 | - | 7 | 832 | ||
| AK017139.1 | - | 7 | 845 | |||
| BC051664.1 | - | 7 | 828 | 30018215 | ||

Serpinb9c
Genomic Map