siRNA Details
| Gene Symbol: Spr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011467.2 | NP_035597.2 | 2 | 369 | ||
| GenBank mRNA | AK154720.1 | - | 373 | |||
| BC010400.1 | - | 2 | 366 | 4219245 | ||
| BC054423.1 | - | 2 | 353 | 4985039 | ||
| CK620382.1 | - | 2 | 337 | |||
| S77493.1 | - | 2 | 371 | |||

Spr
Genomic Map