siRNA Details
| Gene Symbol: Spr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011467.2 | NP_035597.2 | 3 | 790 | ||
| GenBank mRNA | AK004941.1 | - | 3 | 600 | ||
| AK154720.1 | - | 795 | ||||
| AK207238.1 | - | 2 | 195 | |||
| AK216452.1 | - | 2 | 195 | |||
| BC010400.1 | - | 3 | 787 | 4219245 | ||
| BC054423.1 | - | 3 | 774 | 4985039 | ||
| BY590544.1 | - | 1 | 30 | |||
| S77493.1 | - | 3 | 792 | |||
| W65601.1 | - | 2 | 249 | |||

Spr
Genomic Map