siRNA Details
Gene Symbol: Spr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011467.2 | NP_035597.2 | 2 | 378 | ||
GenBank mRNA | AK154720.1 | - | 382 | |||
AK188293.1 | - | 1 | 18 | |||
BC010400.1 | - | 2 | 375 | 4219245 | ||
BC054423.1 | - | 2 | 362 | 4985039 | ||
CK620382.1 | - | 2 | 346 | |||
S77493.1 | - | 2 | 380 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
