siRNA Details
Gene Symbol: Sprr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011471.2 | NP_035601.1 | 2 | 387 | ||
GenBank mRNA | AY158989.1 | - | 2 | 387 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sprr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011471.2 | NP_035601.1 | 2 | 387 | ||
GenBank mRNA | AY158989.1 | - | 2 | 387 |