siRNA Details
| Gene Symbol: Sprr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204427.1 | NP_001191356.1 | 2 | 252 | ||
| NM_011478.2 | NP_035608.1 | 2 | 388 | |||
| GenBank mRNA | BC125629.1 | - | 1 | 282 | 40130144 | |
| BY709657.1 | - | 2 | 388 | |||

Sprr3
Genomic Map