siRNA Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011485.4 | NP_035615.2 | 7 | 1647 | ||
| GenBank mRNA | AK019725.2 | - | 7 | 1687 | ||
| AK054470.1 | - | 7 | 1646 | |||
| BC082283.1 | - | 7 | 1658 | 30436512 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Star| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011485.4 | NP_035615.2 | 7 | 1647 | ||
| GenBank mRNA | AK019725.2 | - | 7 | 1687 | ||
| AK054470.1 | - | 7 | 1646 | |||
| BC082283.1 | - | 7 | 1658 | 30436512 | ||