siRNA Details
| Gene Symbol: Stat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132675.1 | - | 10 | 1011 | ||
| AK157083.1 | - | 9 | 885 | |||
| BC052907.1 | - | 9 | 890 | |||
| BC098499.1 | - | 9 | 878 | 30074608 | ||
| U06923.1 | - | 9 | 884 | |||
| U09351.1 | - | 9 | 1335 | |||

Stat4
Genomic Map