siRNA Details
| Gene Symbol: Stxbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011505.2 | NP_035635.1 | 7 | 754 | ||
| GenBank mRNA | AF152924.1 | - | 7 | 744 | ||
| AK012293.1 | - | 7 | 712 | |||
| AK049001.1 | - | 7 | 723 | |||
| BC032881.1 | - | 7 | 708 | 1224878 | ||

Stxbp4
Genomic Map