siRNA Details
| Gene Symbol: RAB33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004794.2 | NP_004785.1 | 1 | 493 | ||
| GenBank mRNA | AF498959.1 | - | 1 | 229 | ||
| AK094927.1 | - | 1 | 120 | |||
| BC001157.1 | - | 1 | 493 | 3510745 | ||
| BC009996.2 | - | 1 | 493 | 4129324 | ||
| BT007038.1 | - | 1 | 229 | |||
| D14889.1 | - | 1 | 666 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011228, D83279, BC104390, BC104391, AB232628.

RAB33A
Genomic Map