siRNA Details
| Gene Symbol: Plcb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013829.2 | NP_038857.1 | 7 | 520 | ||
| GenBank mRNA | AF332071.1 | - | 7 | 520 | ||
| AF332072.1 | - | 7 | 520 | |||
| AK133360.1 | - | 381 | ||||
| AK148001.1 | - | 8 | 706 | |||
| BC061201.1 | - | 8 | 630 | |||
| BC089521.1 | - | 7 | 483 | |||
| BC129883.1 | - | 7 | 515 | 40110646 | ||

Plcb4
Genomic Map