siRNA Details
| Gene Symbol: Stra13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016665.2 | NP_057874.2 | 5 | 336 | ||
| GenBank mRNA | AK076181.1 | - | 4 | 395 | ||
| AK080500.1 | - | 5 | 308 | |||
| AK170126.1 | - | 5 | 308 | |||
| BC052335.1 | - | 6 | 586 | |||
| CA320983.1 | - | 4 | 275 | |||
| CJ236047.1 | - | 4 | 248 | |||
| U95003.1 | - | 6 | 400 | |||
| U95004.1 | - | 5 | 301 | |||
| U95005.1 | - | 6 | 574 | |||

Stra13
Genomic Map