siRNA Details
| Gene Symbol: Stra13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016665.2 | NP_057874.2 | 5 | 386 | ||
| GenBank mRNA | AK076181.1 | - | 4 | 445 | ||
| AK080500.1 | - | 5 | 358 | |||
| AK170126.1 | - | 5 | 358 | |||
| BC052335.1 | - | 6 | 636 | |||
| CA320983.1 | - | 4 | 325 | |||
| CJ236047.1 | - | 4 | 298 | |||
| U95003.1 | - | 6 | 450 | |||
| U95004.1 | - | 5 | 351 | |||
| U95005.1 | - | 6 | 624 | |||

Stra13
Genomic Map