siRNA Details
| Gene Symbol: TM9SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004800.2 | NP_004791.1 | 1, 2 | 412 | ||
| GenBank mRNA | AK225800.1 | - | 1, 2 | 312 | ||
| AK290514.1 | - | 1, 2 | 367 | |||
| AK315210.1 | - | 1, 2 | 367 | |||
| BC110656.1 | - | 1, 2 | 369 | 3855486 | ||
| BC130293.1 | - | 1, 2 | 246 | 40146324 | ||
| DB024878.1 | - | 1, 2 | 412 | |||
| U81006.1 | - | 1, 2 | 312 | |||

TM9SF2
Genomic Map